missing translation for 'onlineSavingsMsg'
Learn More

Etebenecid, MedChemExpress

Product Code. 16468526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468526 Supplier Medchem Express Supplier No. HYB0939
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Etebenecid is a uricosuric agents, lower uric acid levels in the body by increasing the elimination of uric acid by the kidneys, also inhibits penicillin tubular secretion.

Etebenecid is a uricosuric agents, lower uric acid levels in the body by increasing the elimination of uric acid by the kidneys, also inhibits penicillin tubular secretion.

Specifications

Chemical Name or Material Etebenecid
CAS 1213-06-5
Color White
Purity Grade Notes Research
Molecular Formula C11H15NO4S
Quantity 100 mg
Solubility Information DMSO : 250 mg/mL (971.59 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(S(=O)(N(CC)CC)=O)C=C1
Molecular Weight (g/mol) 257.31
Formula Weight 257.31
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.