missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETEA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530463
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, MEF. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (82%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
Specifications
| ETEA | |
| Polyclonal | |
| Unconjugated | |
| FAF2 | |
| 2210404d11rik; AI462440; ETEA; expressed in T-cells and eosinophils in atopic dermatitis; FAF2; Fas associated factor family member 2; FAS-associated factor 2; Kiaa0887; mKIAA0887; Protein ETEA; protein expressed in T-cells and eosinophils in atopic dermatitis; RGD1306577; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8; UBX8; UBXD8; UBXN3B; Unknown (protein for MGC:129072) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23197, 76577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.48 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q3TDN2, Q96CS3 | |
| FAF2 | |
| Recombinant protein encompassing a sequence within the center region of human ETEA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction