missing translation for 'onlineSavingsMsg'
Learn More

ETC-206, MedChemExpress

Product Code. 16495466
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495466 100 mg 100mg
16474628 10 mg 10mg
16437347 5 mg 5mg
16423176 25 mg 25mg
16420927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495466 Supplier Medchem Express Supplier No. HY112424
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ETC-206 is a selective MNK1 and MNK2 inhibitor with IC50s of 64 nM and 86 nM, respectively.

ETC-206 is a selective MNK1 and MNK2 inhibitor with IC50s of 64 nM and 86 nM, respectively.

Specifications

Chemical Name or Material ETC-206
CAS 1464151-33-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H20N4O2
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (122.41 mM)
SMILES N#CC1=CC=C(C2=CN=C3C=CC(C4=CC=C(C(N5CCOCC5)=O)C=C4)=CN32)C=C1
Molecular Weight (g/mol) 408.45
Formula Weight 408.45
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.