missing translation for 'onlineSavingsMsg'
Learn More

Etazolate, TRC

Product Code. 30375625
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375625 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375625 Supplier Toronto Research Chemicals Supplier No. TRCE8892505MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51022-77-6
Molecular Formula C14 H19 N5 O2
Quantity 5 mg
InChI Formula InChI=1S/C14H19N5O2/c1-5-19-13-10(8-16-19)12(18-17-9(3)4)11(7-15-13)14(20)21-6-2/h7-8H,5-6H2,1-4H3,(H,15,18)
SMILES CCOC(=O)c1cnc2c(cnn2CC)c1NN=C(C)C
IUPAC Name ethyl 1-ethyl-4-(2-propan-2-ylidenehydrazinyl)pyrazolo[3,4-b]pyridine-5-carboxylate
Molecular Weight (g/mol) 289.33
Formula Weight 289.1539
Chemical Name or Material Etazolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.