missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etazolate hydrochloride, Tocris Bioscience™
PDE4 inhibitor
Chemical Identifiers
CAS | 35838-58-5 |
---|---|
Molecular Formula | C14H20ClN5O2 |
Molecular Weight (g/mol) | 325.80 |
MDL Number | MFCD00209848 |
InChI Key | GQJUGJHJUZSJLZ-UHFFFAOYSA-N |
Synonym | etazolate hydrochloride, etazolate hcl, unii-7yo3254y6b, etazolate hydrochloride usan, dsstox_cid_25767, dsstox_rid_81109, dsstox_gsid_45767, 1-ethyl-4-1-methylethylidene hydrazino 1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester hydrochloride, ethyl 1-ethyl-4-2-propan-2-ylidenehydrazinyl pyrazolo 3,4-b pyridine-5-carboxylate hydrochloride |
PubChem CID | 37274 |
IUPAC Name | hydrogen ethyl 1-ethyl-4-[2-(propan-2-ylidene)hydrazin-1-yl]-1H-pyrazolo[3,4-b]pyridine-5-carboxylate chloride |
SMILES | [H+].[Cl-].CCOC(=O)C1=CN=C2N(CC)N=CC2=C1NN=C(C)C |
Description
Phosphodiesterase inhibitor, selective for PDE4 (IC50 = 2.0 μM).Chemical Identifiers
35838-58-5 | |
325.80 | |
GQJUGJHJUZSJLZ-UHFFFAOYSA-N | |
37274 | |
[H+].[Cl-].CCOC(=O)C1=CN=C2N(CC)N=CC2=C1NN=C(C)C |
C14H20ClN5O2 | |
MFCD00209848 | |
etazolate hydrochloride, etazolate hcl, unii-7yo3254y6b, etazolate hydrochloride usan, dsstox_cid_25767, dsstox_rid_81109, dsstox_gsid_45767, 1-ethyl-4-1-methylethylidene hydrazino 1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester hydrochloride, ethyl 1-ethyl-4-2-propan-2-ylidenehydrazinyl pyrazolo 3,4-b pyridine-5-carboxylate hydrochloride | |
hydrogen ethyl 1-ethyl-4-[2-(propan-2-ylidene)hydrazin-1-yl]-1H-pyrazolo[3,4-b]pyridine-5-carboxylate chloride |
Specifications
35838-58-5 | |
MFCD00209848 | |
GQJUGJHJUZSJLZ-UHFFFAOYSA-N | |
hydrogen ethyl 1-ethyl-4-[2-(propan-2-ylidene)hydrazin-1-yl]-1H-pyrazolo[3,4-b]pyridine-5-carboxylate chloride | |
37274 | |
>98% |
C14H20ClN5O2 | |
etazolate hydrochloride, etazolate hcl, unii-7yo3254y6b, etazolate hydrochloride usan, dsstox_cid_25767, dsstox_rid_81109, dsstox_gsid_45767, 1-ethyl-4-1-methylethylidene hydrazino 1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester hydrochloride, ethyl 1-ethyl-4-2-propan-2-ylidenehydrazinyl pyrazolo 3,4-b pyridine-5-carboxylate hydrochloride | |
[H+].[Cl-].CCOC(=O)C1=CN=C2N(CC)N=CC2=C1NN=C(C)C | |
325.80 | |
325.8 | |
Etazolate hydrochloride |
Safety and Handling
Recommended Storage : Store at Room Temperature