missing translation for 'onlineSavingsMsg'
Learn More

Etamivan, MedChemExpress

Product Code. 16478606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478606 Supplier Medchem Express Supplier No. HYB1096
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Etamivan (Ethamivan), an orally active respiratory stimulant, is mainly used in the research of barbiturate overdose and chronic obstructive pulmonary disease.

Etamivan (Ethamivan), an orally active respiratory stimulant, is mainly used in the research of barbiturate overdose and chronic obstructive pulmonary disease.

Specifications

Chemical Name or Material Etamivan
CAS 304-84-7
Color White
Purity Grade Notes Research
Molecular Formula C12H17NO3
Quantity 100 mg
Synonym Ethamivan N,N-Diethylvanillamide
Solubility Information DMSO : ≥ 36 mg/mL (161.24 mM)
SMILES O=C(N(CC)CC)C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol) 223.27
Formula Weight 223.27
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.