missing translation for 'onlineSavingsMsg'
Learn More

Estrone, TRC

Product Code. 30041292
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041292 1 g 1g
30040541 5 g 5g
30009425 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041292 Supplier Toronto Research Chemicals Supplier No. TRCE8890501G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-16-7
Molecular Formula C18 H22 O2
Molecular Weight (g/mol) 270.37
Synonym 3-Hydroxyestra-1,3,5(10)-trien-17-one, Estrone, Estrogens conjugated Imp. I (EP), Estradiol Hemihydrate Imp. A (EP), Estriol Imp. B (EP), Ethinylestradiol Imp. C (EP), (+)-Estrone, (8R,9S,13S,14S)-3-Hydroxy-13-methyl-6,7,8,9,11,12,13,14,15,16-decahydro-17H-cyclopenta[a]phenanthren-17-one, 1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-17-keto-estra-1,3,5-triene, 3-Hydroxyestra-1,3,5(10)-triene-17-one, 3-Hydroxyoestra-1,3,5(10)-trien-17-one, Aquacrine, Crinovaryl, Cristallovar, Crystogen, Destrone, Disynformon, Endofolliculina, Estron, Estrovarin, Estrugenone, Estrusol, Femestrone Inj., Femestrone injection, Femidyn, Fermidyn, Folikrin, Folipex, Folisan, Follestrine, Follestrol, Follicular hormone, Folliculin, Folliculin (steroid), Folliculinum, Follicunodis, Follidrin, Glandubolin, Hiestrone, Hormofollin, Hormovarine, Kestrone, Ketodestrin, Ketohydroxyestrin, Kolpon, Menagen, Menformon, NSC 9699, Oestrin, Oestroform, Oestrone, Oestroperos, Ovifollin, Perlatan, Solliculin, Theelin, Thelestrin, Thelykinin, Thynestron, Tokokin, Unden, WAY 164397, Wynestron, δ1,3,5(10)-Estratrien-3-ol-17-one
IUPAC Name (8R,9S,13S,14S)-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1
Formula Weight 270.162
Chemical Name or Material Estrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.