missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estrone, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Estrone |
| CAS | 53-16-7 |
| Analyte or Component Names | Estrone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Hydroxyestra-1,3,5(10)-trien-17-one, Estrone, Estrogens conjugated Imp. I (EP), Estradiol Hemihydrate Imp. A (EP), Estriol Imp. B (EP), Ethinylestradiol Imp. C (EP), Estradiol Imp. A (EP), (+)-Estrone, (8R,9S,13S,14S)-3-Hydroxy-13-methyl-6,7,8,9,11,12,13,14,15,16-decahydro-17H-cyclopenta[a]phenanthren-17-one, 1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-17-keto-estra-1,3,5-triene, 3-Hydroxyestra-1,3,5(10)-triene-17-one, 3-Hydroxyoestra-1,3,5(10)-trien-17-one, Aquacrine, Crinovaryl, Cristallovar, Crystogen, Destrone, Disynformon, Endofolliculina, Estron, Estrovarin, Estrugenone, Estrusol, Femestrone Inj., Femestrone injection, Femidyn, Fermidyn, Folikrin, Folipex, Folisan, Follestrine, Follestrol, Follicular hormone, Folliculin, Folliculin (steroid), Folliculinum, Follicunodis, Follidrin, Glandubolin, Hiestrone, Hormofollin, Hormovarine, Kestrone, Ketodestrin, Ketohydroxyestrin, Kolpon, Menagen, Menformon, NSC 9699, Oestrin, Oestroform, Oestrone, Oestroperos, Ovifollin, Perlatan, Solliculin, Theelin, Thelestrin, Thelykinin, Thynestron, Tokokin, Unden, WAY 164397, Wynestron, Delta1,3,5(10)-Estratrien-3-ol-17-one, Estradiol Hemihydrate Impurity A, Estriol Impurity B, Estrogens conjugated Impurity I, Ethinylestradiol Impurity C, Estradiol Impurity A |
| InChI Formula | InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction