missing translation for 'onlineSavingsMsg'
Learn More

Estrone b-D-Glucuronide Sodium Salt (>97% by HPLC), TRC

Product Code. 18012330
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012330 10 mg 10mg
18063830 5 mg 5mg
18091830 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18012330 Supplier Toronto Research Chemicals Supplier No. TRCE88905510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15087-01-1
Molecular Formula C24 H29 O8 . Na
Molecular Weight (g/mol) 468.47
Synonym Estrone-B-D-Glucuronide Sodium Salt
IUPAC Name sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[[(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl]oxy]oxane-2-carboxylate
SMILES [Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])ccc34)[C@@H]1CCC2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H30O8.Na/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30;/h3,5,10,14-16,18-21,23,26-28H,2,4,6-9H2,1H3,(H,29,30);/q;+1/p-1/t14-,15-,16+,18+,19+,20-,21+,23-,24+;/m1./s1
Formula Weight 468.176
Percent Purity >95%
Chemical Name or Material Estrone beta-D-Glucuronide sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.