missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estrone 3-Sulfate Sodium Salt, Stabilized with TRIS, TRC
Specifications
Specifications
| Chemical Name or Material | Estrone Sodium Sulfate |
| CAS | 438-67-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H21 O5 S . Na |
| Quantity | 10 mg |
| Synonym | Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, sodium salt (1:1), Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, sodium salt (9CI), Estrone sulfate sodium salt (6CI), Estrone, hydrogen sulfate sodium salt (7CI,8CI), 3-Sulfatoxyestra-1,3,5(10)-trien-17-one sodium salt, Conestoral, Estrone sodium sulfate, Estrone sulfate sodium, Morestin, NSC 18313, Sodium estrone sulfate, Sodium estrone-3-sulfate |
| InChI Formula | InChI=1S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,18+;/m1./s1 |
| SMILES | [Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CCC2=O |
| IUPAC Name | sodium;[(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] sulfate |
| Molecular Weight (g/mol) | 372.41 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?