missing translation for 'onlineSavingsMsg'
Learn More

Estrone 3-Acetate, TRC

Product Code. 30380886
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380886 25 mg
30381958 250 mg
30376394 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380886 Supplier Toronto Research Chemicals Supplier No. TRCE88907525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 901-93-9
Molecular Formula C20 H24 O3
Molecular Weight (g/mol) 312.4
Synonym 3-(Acetyloxy)-estra-1,3,5(10)-trien-17-one, Estrone, acetate (6CI,7CI,8CI), 3-Acetoxy-17-oxo-1,3,5(10)-estratriene, 3-Acetoxyestra-1,3,5(10),15-tetraen-17-one, 3-Acetoxyestra-1,3,5(10)-trien-17-one, 3-Acetoxyestrone, 3-Acetylestrone, 3-O-Acetylestrone, Acetylestrone, Estrone 3-O-acetate, Estrone 3-acetate, Hogival, Oestrone-3-acetate, Puboestrene
IUPAC Name [(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)Oc1ccc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H24O3/c1-12(21)23-14-4-6-15-13(11-14)3-5-17-16(15)9-10-20(2)18(17)7-8-19(20)22/h4,6,11,16-18H,3,5,7-10H2,1-2H3/t16-,17-,18+,20+/m1/s1
Formula Weight 312.1725
Chemical Name or Material Estrone 3-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.