missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estrone 3-Acetate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Estrone 3-Acetate
Specifications
Specifications
| Chemical Name or Material | Estrone 3-Acetate |
| CAS | 901-93-9 |
| Analyte or Component Names | Estrone 3-Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-(Acetyloxy)-estra-1,3,5(10)-trien-17-one, Estrone, acetate (6CI,7CI,8CI), 3-Acetoxy-17-oxo-1,3,5(10)-estratriene, 3-Acetoxyestra-1,3,5(10),15-tetraen-17-one, 3-Acetoxyestra-1,3,5(10)-trien-17-one, 3-Acetoxyestrone, 3-Acetylestrone, 3-O-Acetylestrone, Acetylestrone, Estrone 3-O-acetate, Estrone 3-acetate, Hogival, Oestrone-3-acetate, Puboestrene, Estriol Impurity K |
| InChI Formula | InChI=1S/C20H24O3/c1-12(21)23-14-4-6-15-13(11-14)3-5-17-16(15)9-10-20(2)18(17)7-8-19(20)22/h4,6,11,16-18H,3,5,7-10H2,1-2H3/t16-,17-,18+,20+/m1/s1 |
| SMILES | CC(=O)Oc1ccc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2c1 |
| IUPAC Name | [(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction