missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Estrogen Receptor beta Recombinant Rabbit Monoclonal Antibody (23GB5755), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Estrogen Receptor beta |
|---|---|
| Clone | 23GB5755 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089195
|
Invitrogen™
MA552908 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088540
|
Invitrogen™
MA552909 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Estrogen Receptor beta Recombinant Monoclonal antibody specifically detects Estrogen Receptor beta in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Estrogen Receptor beta | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92731 | |
| 2100 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5755 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor | |
| A synthesized peptide derived from human Estrogen Receptor beta (480-530AA). | |
| Primary | |
| -20°C | |
| ESR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title