Learn More
Estrogen Receptor alpha Monoclonal Antibody (AER314), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512328
Description
MA5-12328 targets Estrogen Receptor in WB, GS, and IP applications and shows reactivity with bovine, human, mouse, rabbit, and rat samples. By Western blot, MA5-12328 detects recombinant ER alpha, but may not be suitable for detection of endogenous protein. The MA5-12328 immunogen is purified, SDS-denatured calf uterus estrogen receptor (ER).
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.
Specifications
| Estrogen Receptor alpha | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P03372, P06211, P19785, P49884 | |
| ESR1 | |
| Purified, SDS-denatured calf uterus estrogen receptor (ER) | |
| 500 μL | |
| Primary | |
| Bovine, Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG1 λ |
| Gel Shift, Immunoprecipitation, Western Blot | |
| AER314 | |
| Unconjugated | |
| ESR1 | |
| DKFZp686N23123; ER; Er alpha; ER36; ERa; ERalpha; ER-alpha; ERalpha protein; ESR; esr 1; Esr1; esr-1; ESRA; Estr; Estra; Estradiol receptor; estrogen nuclear receptor alpha; Estrogen receptor; estrogen receptor 1; estrogen receptor 1 (alpha); estrogen receptor alpha; estrogen receptor alpha c-terminus splice variant 1-2; estrogen receptor alpha E1-E2-1-2; estrogen receptor alpha E1-N2-E2-1-2; estrogen receptor alpha splice variant, CTERP-1; estrogen receptor alpha splice variant, ERalphaDup5; estrogen receptor alpha splice variant, ERalphai45a; estrogen receptor alpha splice variant, ERalphai45bL; estrogen receptor alpha splice variant, ERalphai45bS; estrogen receptor alpha splice variant, ERalphai45c; estrogen receptor alpha splice variant, ERalphai56; estrogen receptor alpha splice variant, ERalphai67; estrogen receptor alpha variant delta 4; estrogen receptor protein; estrogen receptor, alpha; ESTRR; hER-alpha36; I79_001166; Nr3a1; nuclear receptor; nuclear receptor subfamily 3 group A member 1; RNESTROR; RP1-130E4.1; unnamed protein product | |
| Mouse | |
| Protein G | |
| RUO | |
| 100351261, 13982, 2099, 24890, 407238 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.