Learn More
Estrogen Receptor alpha Monoclonal Antibody (6F11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513304
Description
MA5-13304 has successfully been used in IHC, IP, and Western blot procedures. Paraffin sections require an antigen retrieval pretreatment with sodium citrate buffer (pH 6.0) prior to staining. Use Samboni’s fixative for optimal staining results. Positive control of Breast carcinoma suggested.
Estrogen Receptor alpha Monoclonal antibody specifically detects Estrogen Receptor alpha in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
| Estrogen Receptor alpha | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.09% sodium azide | |
| P03372, P06211 | |
| ESR1 | |
| Recombinant estrogen receptor protein (alpha form) | |
| RUO | |
| 2099, 24890 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 6F11 | |
| Unconjugated | |
| ESR1 | |
| DKFZp686N23123; ER; Er alpha; ER36; ERa; ERalpha; ER-alpha; ERalpha protein; ESR; esr 1; Esr1; esr-1; ESRA; Estr; Estra; Estradiol receptor; estrogen nuclear receptor alpha; Estrogen receptor; estrogen receptor 1; estrogen receptor 1 (alpha); estrogen receptor alpha; estrogen receptor alpha c-terminus splice variant 1-2; estrogen receptor alpha E1-E2-1-2; estrogen receptor alpha E1-N2-E2-1-2; estrogen receptor alpha splice variant, CTERP-1; estrogen receptor alpha splice variant, ERalphaDup5; estrogen receptor alpha splice variant, ERalphai45a; estrogen receptor alpha splice variant, ERalphai45bL; estrogen receptor alpha splice variant, ERalphai45bS; estrogen receptor alpha splice variant, ERalphai45c; estrogen receptor alpha splice variant, ERalphai56; estrogen receptor alpha splice variant, ERalphai67; estrogen receptor alpha variant delta 4; estrogen receptor protein; estrogen receptor, alpha; ESTRR; hER-alpha36; I79_001166; Nr3a1; nuclear receptor; nuclear receptor subfamily 3 group A member 1; RNESTROR; RP1-130E4.1; unnamed protein product | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.