missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Estriol Monoclonal Antibody (6H8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549700
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Estriol is the main estrogen in the blood and urine of pregnant females. Serum estriol determinations reflect the status of the fetus and are used to evaluate fetal distress and placental function.
Specifications
| Estriol | |
| Monoclonal | |
| 0.993 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 6H8 | |
| Unconjugated | |
| C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion | |
| Conjugate of E3 hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction