missing translation for 'onlineSavingsMsg'
Learn More

Estriol Methyl Ether, TRC

Product Code. 30383374
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383374 100 mg -
30374179 250 mg -
30380134 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383374 Supplier Toronto Research Chemicals Supplier No. TRCE668660100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1474-53-9
Molecular Formula C19 H26 O3
Molecular Weight (g/mol) 302.41
Synonym 3-Methoxyestra-1,3,5(10)-triene-16alpha,17beta-diol, Estriol Imp. C (EP), Estra-1,3,5(10)-triene-16,17-diol, 3-methoxy-, (16α,17β)-, Estra-1,3,5(10)-triene-16α,17β-diol, 3-methoxy- (6CI,7CI,8CI), (16α,17β)-3-Methoxyestra-1,3,5(10)-triene-16,17-diol, 13β-Methyl-3-methoxygona-1,3,5(10)triene-16α,17β-diol, 16α,17β-Estriol-3-methyl ether, 3-Methoxyestriol, E 2710, Estriol 3-methyl ether, Estriol methyl ether
IUPAC Name (8R,9S,13S,14S,16R,17R)-3-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-16,17-diol
SMILES COc1ccc2[C@H]3CC[C@]4(C)[C@@H](O)[C@H](O)C[C@H]4[C@@H]3CCc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H26O3/c1-19-8-7-14-13-6-4-12(22-2)9-11(13)3-5-15(14)16(19)10-17(20)18(19)21/h4,6,9,14-18,20-21H,3,5,7-8,10H2,1-2H3/t14-,15-,16+,17-,18+,19+/m1/s1
Formula Weight 302.1882
Chemical Name or Material 3-Methoxyestra-1,3,5(10)-triene-16alpha,17beta-diol (Estriol 3-Methyl Ether)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.