missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estriol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Estriol |
| CAS | 50-27-1 |
| Analyte or Component Names | Estriol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H24 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Estriol, Aacifemine, Colpogyn, Destriol, Estratriol, Follicular hormone hydrate, Gynasan, Holin V, Hormomed, Incurin, Klimax E, Klimoral, NSC 12169, OE3, Oekolp, Oestriol, Ortho-Gynest, Ovesterin, Ovestin, Ovo-Vinces, Theelol, Tridestrin, Trihydroxyestrin, Triovex, (16alpha,17beta)-Estra-1,3,5(10)-triene-3,16,17-triol, 13beta-Methyl-1,3,5(10)-gonatriene-3,16alpha,17beta-triol, 16alpha,17beta-Estriol, 16alpha-Estriol, 16alpha-Hydroxyestradiol, 3,16alpha,17beta-Estriol, 3,16alpha,17beta-Trihydroxyestra-1,3,5(10)-triene, Estra-1,3,5(10)-triene-3,16alpha,17beta-triol |
| InChI Formula | InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1 |
| Solution Type | Single Solution |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction