missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Estramustine phosphate sodium
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 250 mg |
| Solubility | Solubility aq. soln.: 50 g/L |
| Formula Weight | 564.35 |
| InChI Key | IIUMCNJTGSMNRO-VVSKJQCTSA-L |
| SMILES | [Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O |
| IUPAC Name | disodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate |
| Molecular Weight (g/mol) | 564.35 |
| Chemical Name or Material | Estramustine phosphate sodium |
Safety and Handling
Signal Word
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?