missing translation for 'onlineSavingsMsg'
Learn More

Estramustine Disodium Phosphate (~85%), TRC

Product Code. 30386595
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386595 50 mg
30372278 100 mg
30377014 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386595 Supplier Toronto Research Chemicals Supplier No. TRCE88898350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52205-73-9
Molecular Formula C23H30Cl2NNa2O6P
Molecular Weight (g/mol) 564.35
Synonym Emcyt, Estracyt, Estramustine Phosphate Disodium, Estramustine Phosphate Disodium Salt, Estramustine Phosphate Sodium, Ro 21-8837/001
IUPAC Name disodium;[(8R,9S,13S,14S,17S)-3-[bis(2-chloroethyl)carbamoyloxy]-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] phosphate
SMILES [Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)N(CCCl)CCCl)ccc34)[C@@H]1CC[C@@H]2OP(=O)([O-])[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H32Cl2NO6P.2Na/c1-23-9-8-18-17-5-3-16(31-22(27)26(12-10-24)13-11-25)14-15(17)2-4-19(18)20(23)6-7-21(23)32-33(28,29)30;;/h3,5,14,18-21H,2,4,6-13H2,1H3,(H2,28,29,30);;/q;2*+1/p-2/t18-,19-,20+,21+,23+;;/m1../s1
Formula Weight 563.0983
Chemical Name or Material Estramustine Disodium Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.