missing translation for 'onlineSavingsMsg'
Learn More

Estragole, Mikromol™

Product Code. 17951976
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951976 Supplier Mikromol Supplier No. MM0164.02
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Estragole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estragole
CAS 140-67-0
Analyte or Component Names Estragole
Shipping Condition Room Temperature
Molecular Formula C10 H12 O
Certifications/Compliance ISO 17025
Synonym Estragole, Benzene, 1-methoxy-4-(2-propen-1-yl)-, Anisole, p-allyl- (8CI), Benzene, 1-methoxy-4-(2-propenyl)- (9CI), Estragole (6CI), Ether, p-allylphenyl methyl (7CI), 1-Allyl-4-methoxybenzene, 1-Methoxy-4-(2-propen-1-yl)benzene, 1-Methoxy-4-(2-propenyl)benzene, 3-(4-Methoxyphenyl)-1-propene, 3-(4-Methoxyphenyl)propene, 3-(p-Methoxyphenyl)-1-propene, 3-(p-Methoxyphenyl)propene, 4-Allylanisole, 4-Allylmethoxybenzene, 4-Methoxyallylbenzene, AUSTL 21320, Chavicol methyl ether, Chavicol, O-methyl-, Chavicol, methyl-, Esdragol, Esdragole, Esdragon, Esteragol, Estragenole, Estragol, Isoanethole, Methyl chavicol, NSC 404113, O-Methylchavicol, p-Allylanisole, p-Allylphenyl methyl ether, p-Methoxyallylbenzene
InChI Formula InChI=1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3,5-8H,1,4H2,2H3
SMILES COc1ccc(CC=C)cc1
IUPAC Name 1-methoxy-4-prop-2-enylbenzene
Molecular Weight (g/mol) 148.2
Quantity 100 mg
Formula Weight 148.0888 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.