missing translation for 'onlineSavingsMsg'
Learn More

β-Estradiol, MP Biomedicals™

Product Code. 11428030 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428030 100 g 100g
11408030 5 g 5g
11418030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428030 Supplier MP Biomedicals Supplier No. 0210165680
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.388
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

Quantity 100 g
Formula Weight 272.4
Physical Form Solid
Chemical Name or Material β-Estradiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.