missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Estradiol Monoclonal Antibody (AB4.2)
GREENER_CHOICE

Product Code. 12867972
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12867972 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12867972 Supplier Invitrogen™ Supplier No. MIE0106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIE0106 targets Estradiol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIE0106 immunogen is estradiol-6-CMO-KLH. MIE0106 was formerly sold as a Seradyn product.

Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Estradiol
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone AB4.2
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 17-Beta-Estradiol; 17-beta-HSD
Host Species Mouse
Immunogen Estradiol-6-CMO-KLH.
Purification Method Ion-exchange chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.