Learn More
Estradiol Monoclonal Antibody (4S112 (BGN/06/88112)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183370
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-83370 exhibits negligible cross-reactivity with estriol and testosterone. Mouse anti 17-beta-Estradiol antibody, clone 4S112 (BGN/06/88112) recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle.
Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.Specifications
Estradiol | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA | |
4S112 (BGN/06/88112) | |
Unconjugated | |
17-Beta-Estradiol; 17-beta-HSD | |
17-beta-estradiol conugated to bovine serum albumin at position 6. | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |