missing translation for 'onlineSavingsMsg'
Learn More

Estradiol Acetate, TRC

Product Code. 30401994
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401994 50 mg -
30407885 500 mg -
30406711 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401994 Supplier Toronto Research Chemicals Supplier No. TRCH45414050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4245-41-4
Molecular Formula C20H26O3
Molecular Weight (g/mol) 314.42
Synonym (8R,9S,13S,14S,17S)-17-Hydroxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthren-3-yl acetate, 17beta-Estradiol 3-acetate, 3-Acetoxyestra-1,3,5(10)-trien-17beta-ol, 3-O-Acetylestradiol, E 3A, Estra-1,3,5(10)-triene-3,17beta-diyl 3-acetate, Estradiol-3-acetate, Menoring, beta-Estradiol 3-acetate, 17beta-Hydroxy-3-acetoxyestra-1,3,5(10)-triene
IUPAC Name [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)Oc1ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H26O3/c1-12(21)23-14-4-6-15-13(11-14)3-5-17-16(15)9-10-20(2)18(17)7-8-19(20)22/h4,6,11,16-19,22H,3,5,7-10H2,1-2H3/t16-,17-,18+,19+,20+/m1/s1
Formula Weight 314.1882
Chemical Name or Material Estradiol Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.