missing translation for 'onlineSavingsMsg'
Learn More

Estradiol 17-Valerate-d9, TRC

Product Code. 30388450
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388450 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30388450 Supplier Toronto Research Chemicals Supplier No. TRCE8888621MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estradiol 17-Valerate-d9
CAS 1316648-98-2
Molecular Formula C23H23D9O3
Quantity 1 mg
Synonym (17β)-Estra-1,3,5(10)-triene-3,17-diol 17-Pentanoate-d9, Estradiol Valerate-d9, 3-Hydroxy-17β-(valeroyl-d9)oxyestra-1,3,5(10)-triene, Atladiol-d9, Climaval-d9, Deladiol, -d9 Delestrogen-d9, Delestrogen 4x-d9, Estradiol 17β-Valerate-d9, Estradiol Valerianate-d9, Estraval-d9, Femogex-d9, Gynokadin-d9, NSC 17590-d9, Neofollin-d9, Nuvelle-d9, Progynova-d9, Valergen-d9
InChI Formula InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1/i1D3,3D2,4D2,5D2
SMILES OC1=CC(CC[C@@]([H])([C@@]23[H])[C@]4([H])CC[C@@]2([C@@H](OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)CC3)C)=C4C=C1
Molecular Weight (g/mol) 365.55
Formula Weight 365.2916
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.