missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Estradiene dione

Product Code. 16574244 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16574244

Brand: Thermo Scientific Chemicals 463770010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5571-36-8
Molecular Formula C20H26O3
Molecular Weight (g/mol) 314.43
InChI Key XUOQKQRMICQUQC-UHFFFAOYNA-N
IUPAC Name 11a-methyl-1,2,3,3a,3b,4,5,6,8,9,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-7,2'-[1,3]dioxolan]-1-one
SMILES CC12CC=C3C(CCC4=C3CCC3(C4)OCCO3)C1CCC2=O

Specifications

Melting Point 156.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in hot ethanol
Formula Weight 314.43
Chemical Name or Material Estradiene dione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.