missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRG Polyclonal Antibody

ESRRG Polyclonal Antibody
Brand: Invitrogen PA533524
This item is not returnable.
View return policy
Description
ESRRG Polyclonal Antibody for IHC (P)
Specifications
| ESRRG | |
| Polyclonal | |
| Unconjugated | |
| Esrrg | |
| DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100050530, 100355568, 100622056, 100760533, 2104, 26381, 360896, 421357, 478961, 536732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P62508, P62509, P62510 | |
| Esrrg | |
| Synthetic 15 amino acid peptide from C-terminus of human ESRRG / ERR3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction