missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ESRRG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533525
This item is not returnable.
View return policy
Description
ESRRG Polyclonal Antibody for IHC (P)
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.
Specifications
| ESRRG | |
| Polyclonal | |
| Unconjugated | |
| Esrrg | |
| DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100050530, 100355568, 100760533, 2104, 26381, 360896, 478961, 536732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P62508, P62509, P62510 | |
| Esrrg | |
| Synthetic 14 amino acid peptide from N-terminus of human ESRRG / ERR3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction