Learn More
Invitrogen™ ESRP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113717
Description
Immunogen sequence: ATAGALGRDL GSDETDLILL VWQVVEPRSR QVGTLHKSLV RAEAAALSTQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing program induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.
Specifications
| ESRP2 | |
| Polyclonal | |
| Unconjugated | |
| ESRP2 | |
| 9530027K23Rik; Epithelial splicing regulatory protein 2; ESRP2; PP7059; RBM35B; RGD1310855; RNA binding motif protein 35A; RNA binding motif protein 35B; RNA-binding motif protein 35B; RNA-binding protein 35B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80004 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H6T0 | |
| ESRP2 | |
| Synthetic peptide directed towards the N terminal region of human RBM35B (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.