missing translation for 'onlineSavingsMsg'
Learn More

ESRP2 Antibody (22C5.H7.A6), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18335371 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335371 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335371 Supplier Novus Biologicals Supplier No. NBP177972AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ESRP2 Monoclonal specifically detects ESRP2 in Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 22C5.H7.A6
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias epithelial splicing regulatory protein 2, FLJ21918, FLJ22248, RBM35BRNA-binding protein 35B, RNA binding motif protein 35A, RNA binding motif protein 35B, RNA-binding motif protein 35B
Host Species Mouse
Immunogen ESRP2 Antibody (22C5.H7.A6) is produced by repeated immunizations of full length recombinant mouse Esrp-2 fusion protein. (Uniprot: Q8K0G8)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80004
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.