missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21045-1-AP-20UL
This item is not returnable.
View return policy
Description
RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5′ UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.Specifications
| ESRP1 | |
| Polyclonal | |
| Unconjugated | |
| ESRP1 | |
| ESRP1, RBM35A, RNA binding motif protein 35A, RNA binding protein 35A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54845 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6NXG1 | |
| ESRP1 | |
| ESRP1 Fusion Protein Ag15386 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction