missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ESRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525833
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
Specifications
| ESRP1 | |
| Polyclonal | |
| Unconjugated | |
| ESRP1 | |
| 2210008M09Rik; A630065D16; BC031468; epithelial splicing regulatory protein 1; Esrp1; Rbm35a; RGD1560481; RMB35A; RNA binding motif protein 35A; RNA-binding motif protein 35A; RNA-binding protein 35A | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 54845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6NXG1 | |
| ESRP1 | |
| KLH conjugated synthetic peptide between 182-211 amino acids of human ESRP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction