missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ESRP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849252
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849252 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849252 Supplier Invitrogen™ Supplier No. PA521109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat colon lysate. PA5-21109 can be used with blocking peptide PEP-1223.

RBM35A, also known as ESRP1, is a mRNA splicing factor that with its related protein RBM35B (ESRP2) are coordinators of an epithelial cell-type-specific splicing program. RBM35A contains three putative RNA recognition motifs and acts by directly binding specific sequences in mRNAs. RBM35A is involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs. Other recent studies have shown that RMB35A may also act as a novel tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ESRP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ESRP1
Gene Accession No. B2RYD2, Q3US41, Q6NXG1
Gene Alias 2210008M09Rik; A630065D16; BC031468; epithelial splicing regulatory protein 1; Esrp1; Rbm35a; RGD1560481; RMB35A; RNA binding motif protein 35A; RNA-binding motif protein 35A; RNA-binding protein 35A
Gene Symbols ESRP1
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human RBM35A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207920, 500409, 54845
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.