missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76536
This item is not returnable.
View return policy
Description
ESRP1 Polyclonal specifically detects ESRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ESRP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| epithelial splicing regulatory protein 1, FLJ20171, RBM35A, RMB35A, RNA binding motif protein 35A, RNA-binding motif protein 35A, RNA-binding protein 35A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54845 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
| NP_001030087 | |
| ESRP1 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of human RBM35A. The immunogen is located within amino acids 30 - 80 of RBM35A. Amino Acid Squence: LVRPDQLELTED | |
| 0.1 mg | |
| Primary | |
| RBM35A antibody is predicted to not cross-react with other RBM35/ESRP family members. At least five isoforms of RBM35A are known to exist; this antibody will detect all five. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction