missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ESRP1/2 Polyclonal specifically detects ESRP1/2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ESRP1/2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | epithelial splicing regulatory protein 2,9530027K23Rik, Rbm35b, RNA binding motif protein 35b, RNA-binding motif protein 35B, RNA-binding protein 35B |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human ESRP1/2 |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?