missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR2 Beta-3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ESR2 Beta-3 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16876905
|
Proteintech
55441-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16866905
|
Proteintech
55441-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The human ER-beta is a newly discovered estrogen receptor initially cloned and characterized from testis. The size and structure of ER-beta is very similar to ER-alpha with the ligand and DNA binding domains being highly conserved, while the amino terminus which serves as their transactivation domain has diverged significantly. Similar in function to ER-alpha ER-beta binds to estrogen with a high affinity and regulates estrogen specific gene activation through direct interaction with estrogen response elements (ERE′s).Specifications
| ESR2 Beta-3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92731 | |
| 2100 | |
| Peptide | |
| Primary | |
| -20°C | |
| ESR2 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2 | |
| ESR2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title