missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESR1 Mouse anti-Human, Rat, Clone:6A6E8, Abnova™
Mouse monoclonal antibody raised against recombinant human ESR1.
Brand: Abnova MAB17277.100ug
This item is not returnable.
View return policy
Description
This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5′ UTRs and use different promoters. [provided by RefSeq]Specifications
| ESR1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| ESR1 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 6A6E8 | |
| estrogen receptor 1 | |
| In PBS (0.05% sodium azide) | |
| DKFZp686N23123, ER, ESR, ESRA, Era, NR3A1 | |
| Recombinant protein corresponding to amino acid 2-185 of human ESR1 from E. coli. | |
| 2099 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction