missing translation for 'onlineSavingsMsg'
Learn More

Esprocarb 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30325822
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30325822

Brand: Dr. Ehrenstorfer DREXA13212000AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Esprocarb
CAS 85785-20-2
Analyte or Component Names Esprocarb
Shipping Condition Room Temperature
Molecular Formula C15 H23 N O S
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
InChI Formula InChI=1S/C15H23NOS/c1-5-16(13(4)12(2)3)15(17)18-11-14-9-7-6-8-10-14/h6-10,12-13H,5,11H2,1-4H3
Solution Type Single Solution
SMILES CCN(C(C)C(C)C)C(=O)SCc1ccccc1
IUPAC Name S-benzyl N-ethyl-N-(3-methylbutan-2-yl)carbamothioate
Solvent or Matrix Acetone
Industry Type Environmental Testing
Molecular Weight (g/mol) 265.41
Quantity 1 mL
Formula Weight 265.15 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.