missing translation for 'onlineSavingsMsg'
Learn More

Esonarimod, MedChemExpress

Product Code. 16487768
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487768 10 mg 10mg
16487616 100 mg 100mg
16443737 50 mg 50mg
16430238 5 mg 5mg
16401819 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487768 Supplier Medchem Express Supplier No. HY19440
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Esonarimod (KE-298) is an antirheumatic agent.

Esonarimod (KE-298) is an antirheumatic agent.

Specifications

Chemical Name or Material Esonarimod
Physical Form Solid
CAS 101973-77-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H16O4S
For Use With (Application) COVID-19-immunoregulation
Synonym KE-298
Solubility Information DMSO : 100 mg/mL (356.71 mM; Need ultrasonic)
SMILES CC1=CC=C(C(CC(CSC(C)=O)C(O)=O)=O)C=C1
Molecular Weight (g/mol) 280.34
Formula Weight 280.34
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.