missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esomeprazole Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Esomeprazole Sodium
Specifications
Specifications
| Chemical Name or Material | Esomeprazole Sodium |
| CAS | 161796-78-7 |
| Analyte or Component Names | Esomeprazole Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 N3 O3 S . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Esomeprazole sodium salt, (S)-(-)-Omeprazole Sodium, (S)-Omeprazole Sodium Salt, Esomeprazole Sodium |
| InChI Formula | InChI=1S/C17H18N3O3S.Na/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;/h5-8H,9H2,1-4H3;/q-1;+1/t24-;/m0./s1 |
| SMILES | [Na+].COc1ccc2[n-]c(nc2c1)[S@@](=O)Cc3ncc(C)c(OC)c3C |
| IUPAC Name | sodium;5-methoxy-2-[(S)-(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]benzimidazol-1-ide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction