missing translation for 'onlineSavingsMsg'
Learn More

Esomeprazole sodium, MedChemExpress

Product Code. 16453287
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16453287

Brand: Medchem Express HY17023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Esomeprazole sodium ((S)-Omeprazole sodium) is a potent and orally active proton pump inhibitor. Esomeprazole reduces acid secretion through inhibition of the H+, K+-ATPase in gastric parietal cells. Esomeprazole acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases. Esomeprazole has the potential for symptomatic gastroesophageal reflux disease research.

Esomeprazole sodium ((S)-Omeprazole sodium) is a potent and orally active proton pump inhibitor. Esomeprazole reduces acid secretion through inhibition of the H+, K+-ATPase in gastric parietal cells. Esomeprazole acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases. Esomeprazole has the potential for symptomatic gastroesophageal reflux disease research.

Specifications

Chemical Name or Material Esomeprazole sodium
CAS 161796-78-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18N3NaO3S
Quantity 50 mg
Synonym (S)-Omeprazole sodium(-)-Omeprazole sodium
Solubility Information DMSO : 100 mg/mL (272.18 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (136.09 mM; Need ultrasonic)
SMILES O=[S@](C1=NC2=CC(OC)=CC=C2N1[Na])CC3=NC=C(C)C(OC)=C3C
Molecular Weight (g/mol) 367.4
Formula Weight 367.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.