missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esomeprazole magnesium trihydrate, MedChemExpress
Description
Esomeprazole magnesium trihydrate ((S)-Omeprazole magnesium trihydrate) is a potent and orally active H+, K+-ATPase inhibitor. Esomeprazole magnesium trihydrate has the potential for upper intestinal disorders and gastroesophageal reflux disease research. Esomeprazole magnesium trihydrate acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases.
Specifications
Specifications
| Chemical Name or Material | Esomeprazole magnesium trihydrate |
| CAS | 217087-09-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H42MgN6O9S2 |
| Quantity | 50 mg |
| Synonym | (S)-Omeprazole magnesium trihydrate(-)-Omeprazole magnesium trihydrate |
| Solubility Information | DMSO : 50 mg/mL (65.17 mM; Need ultrasonic) |
| SMILES | [H]O[H].[H]O[H].[H]O[H].COC1=CC=C2[N]3=C([N-]C2=C1)[S](CC(N=CC(C)=C4OC)=C4C)=[O][Mg+2]35[N](C6=CC=C7OC)=C([N-]C6=C7)[S](CC(N=CC(C)=C8OC)=C8C)=[O]5 |
| Molecular Weight (g/mol) | 767.17 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?