missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esomeprazole magnesium trihydrate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
Esomeprazole magnesium trihydrate ((S)-Omeprazole magnesium trihydrate) is a potent and orally active H+, K+-ATPase inhibitor. Esomeprazole magnesium trihydrate has the potential for upper intestinal disorders and gastroesophageal reflux disease research. Esomeprazole magnesium trihydrate acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases.
Specifications
Specifications
| Chemical Name or Material | Esomeprazole magnesium trihydrate |
| CAS | 217087-09-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H42MgN6O9S2 |
| Quantity | 100 mg |
| Synonym | (S)-Omeprazole magnesium trihydrate(-)-Omeprazole magnesium trihydrate |
| Solubility Information | DMSO : 50 mg/mL (65.17 mM; Need ultrasonic) |
| SMILES | [H]O[H].[H]O[H].[H]O[H].COC1=CC=C2[N]3=C([N-]C2=C1)[S](CC(N=CC(C)=C4OC)=C4C)=[O][Mg+2]35[N](C6=CC=C7OC)=C([N-]C6=C7)[S](CC(N=CC(C)=C8OC)=C8C)=[O]5 |
| Molecular Weight (g/mol) | 767.17 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction