missing translation for 'onlineSavingsMsg'
Learn More

Esmolol Acid, TRC

Product Code. 17964025
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964025 250 mg 250mg
17943685 10 mg 10mg
17932345 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964025 Supplier Toronto Research Chemicals Supplier No. TRCE668010250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81148-15-4
Molecular Formula C15 H23 N O4
Molecular Weight (g/mol) 281.35
IUPAC Name 3-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]propanoic acid
SMILES CC(C)NCC(O)COc1ccc(CCC(=O)O)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H23NO4/c1-11(2)16-9-13(17)10-20-14-6-3-12(4-7-14)5-8-15(18)19/h3-4,6-7,11,13,16-17H,5,8-10H2,1-2H3,(H,18,19)
Formula Weight 281.1627
Percent Purity >95%
Chemical Name or Material Esmolol Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.