missing translation for 'onlineSavingsMsg'
Learn More

Eslicarbazepine Acetate, TRC

Product Code. 30385579
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385579 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385579 Supplier Toronto Research Chemicals Supplier No. TRCE65900050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eslicarbazepine Acetate
CAS 236395-14-5
Molecular Formula C17 H16 N2 O3
Quantity 50 mg
Synonym (10S)-10-(Acetyloxy)-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide, (S)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide, BIA 2-093, Eslicarbazepine acetate, Exelief, Sep 0002093
InChI Formula InChI=1S/C17H16N2O3/c1-11(20)22-16-10-12-6-2-4-8-14(12)19(17(18)21)15-9-5-3-7-13(15)16/h2-9,16H,10H2,1H3,(H2,18,21)/t16-/m0/s1
SMILES CC(=O)O[C@H]1Cc2ccccc2N(C(=O)N)c3ccccc13
IUPAC Name [(5S)-11-carbamoyl-5,6-dihydrobenzo[b][1]benzazepin-5-yl] acetate
Molecular Weight (g/mol) 296.32
Formula Weight 296.1161
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.