missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ESF1 Polyclonal antibody specifically detects ESF1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ESF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ABT1-associated protein, ABTAP, bA526K24.1, C20orf6, chromosome 20 open reading frame 6, ESF1 homolog, ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae), FLJ20368 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 801 and 851 of human ABT1-Associated Protein using the numbering given in entry NP_057733.2 (GeneID 51575). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?