missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15244-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a serine hydrolase that belongs to the esterase D family. The encoded enzyme is active toward numerous substrates including O-acetylated sialic acids, and it may be involved in the recycling of sialic acids. This gene is used as a genetic marker for retinoblastoma and Wilson′s disease.Specifications
| ESD | |
| Polyclonal | |
| Unconjugated | |
| ESD | |
| ESD, Esterase D, FGH, S formylglutathione hydrolase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13885, 2098, 290401 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0BNE5, P10768, Q9R0P3 | |
| Esd | |
| ESD Fusion Protein Ag7457 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction