Learn More
ESD Clean Room Chair Black
Product Code. 15424593
Encompass_Preferred
Product Code. 15424593 Supplier Techsit Supplier No. WS1720RR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Lab chairs certified for class-5 clean rooms according to DIN EN ISO 14644-1 standard. All materials are washable and disinfectant resistant with the vinyl being blood, sweat and urine resistant in accordance with the DIN 53160 standard.
  • Black vinyl backrest and seat.
  • Chrome-plated cover tube.
  • Height adjustment with safety gas spring.
  • Polished diecast aluminum five-star base with conductive glides.