missing translation for 'onlineSavingsMsg'
Learn More

Esculin Sesquihydrate, TRC

Product Code. 30384305
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384305 100 mg
30371834 50 mg
30374331 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384305 Supplier Toronto Research Chemicals Supplier No. TRCE658958100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66778-17-4
Molecular Formula 2 C15 H16 O9 . 3 H2 O
Molecular Weight (g/mol) 734.61
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
SMILES O.O.O.OC[C@H]1O[C@@H](Oc2cc3C=CC(=O)Oc3cc2O)[C@H](O)[C@@H](O)[C@@H]1O.OC[C@H]4O[C@@H](Oc5cc6C=CC(=O)Oc6cc5O)[C@H](O)[C@@H](O)[C@@H]4O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C15H16O9.3H2O/c2*16-5-10-12(19)13(20)14(21)15(24-10)23-9-3-6-1-2-11(18)22-8(6)4-7(9)17;;;/h2*1-4,10,12-17,19-21H,5H2;3*1H2/t2*10-,12-,13+,14-,15-;;;/m11.../s1
Formula Weight 734.191
Chemical Name or Material Esculin Sesquihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.